missing translation for 'onlineSavingsMsg'
Learn More

4-Styrylpyridine 98.0+%, TCI America™

Catalog No. S04751G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04751G 1 g
S04755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S04751G Supplier TCI America Supplier No. S04751G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103-31-1
Molecular Formula C13H11N
Molecular Weight (g/mol) 181.24
MDL Number MFCD00137749
InChI Key QKHRGPYNTXRMSL-VOTSOKGWSA-N
Synonym 4-Stilbazole
PubChem CID 641098
IUPAC Name 4-[(1E)-2-phenylethenyl]pyridine
SMILES C(=C/C1=CC=NC=C1)\C1=CC=CC=C1

Specifications

Melting Point 131°C
Quantity 1 g
Formula Weight 181.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Styrylpyridine