missing translation for 'onlineSavingsMsg'
Learn More

4-Quinolinol 98.0+%, TCI America™

Supplier:  TCI America Q002925G

 View more versions of this product

Catalog No. Q002925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Quinolinol
203°C
C9H7NO
25 g
PMZDQRJGMBOQBF-UHFFFAOYSA-N
1,4-dihydroquinolin-4-one
69141
145.16
Crystalline Powder
611-36-9
White-Yellow
MFCD00006777,MFCD00956391
4-hydroxyquinoline, quinolin-4-ol, 4-quinolinol, quinolin-4 1h-one, 4 1h-quinolinone, kynurine, 4-hydroxy quinoline, quinoline, 4-hydroxy, ccris 4329, 4-chinolinol
O=C1C=CNC2=CC=CC=C12
145.16
CHEBI:15815
≥98.0% (GC,T)

Chemical Identifiers

611-36-9
145.16
PMZDQRJGMBOQBF-UHFFFAOYSA-N
69141
1,4-dihydroquinolin-4-one
C9H7NO
MFCD00006777,MFCD00956391
4-hydroxyquinoline, quinolin-4-ol, 4-quinolinol, quinolin-4 1h-one, 4 1h-quinolinone, kynurine, 4-hydroxy quinoline, quinoline, 4-hydroxy, ccris 4329, 4-chinolinol
CHEBI:15815
O=C1C=CNC2=CC=CC=C12
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : VC4070000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions