missing translation for 'onlineSavingsMsg'
Learn More

4-Quinolinol 98.0+%, TCI America™

Catalog No. p-7135626
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00295G 5 g
Q002925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q00295G Supplier TCI America Supplier No. Q00295G
Only null left

Chemical Identifiers

CAS 611-36-9
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00006777,MFCD00956391
InChI Key PMZDQRJGMBOQBF-UHFFFAOYSA-N
Synonym 4-hydroxyquinoline, quinolin-4-ol, 4-quinolinol, quinolin-4 1h-one, 4 1h-quinolinone, kynurine, 4-hydroxy quinoline, quinoline, 4-hydroxy, ccris 4329, 4-chinolinol
PubChem CID 69141
ChEBI CHEBI:15815
IUPAC Name 1,4-dihydroquinolin-4-one
SMILES O=C1C=CNC2=CC=CC=C12

Specifications

Melting Point 203°C
Color White-Yellow
Quantity 5 g
Formula Weight 145.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Quinolinol
RTECSNumber VC4070000
TSCA No