missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pyrrolidinopyridine 98.0+%, TCI America™
$76.43 - $221.83
Chemical Identifiers
| CAS | 2456-81-7 |
|---|---|
| Molecular Formula | C9H12N2 |
| Molecular Weight (g/mol) | 148.209 |
| MDL Number | MFCD00006415 |
| InChI Key | RGUKYNXWOWSRET-UHFFFAOYSA-N |
| Synonym | 4-pyrrolidinopyridine, 4-pyrrolidin-1-yl pyridine, 4-1-pyrrolidinyl pyridine, pyridine, 4-1-pyrrolidinyl, 4-pyrrolidin-1-yl-pyridine, 4-pyrrolidinylpyridine, 1-4-pyridyl pyrrolidine, zlchem 148, 4-pyrrolidinepyridine, 4-pyrrollidinopyridine |
| PubChem CID | 75567 |
| IUPAC Name | 4-pyrrolidin-1-ylpyridine |
| SMILES | C1CCN(C1)C2=CC=NC=C2 |
Chemical Identifiers
| 2456-81-7 | |
| 148.209 | |
| RGUKYNXWOWSRET-UHFFFAOYSA-N | |
| 75567 | |
| C1CCN(C1)C2=CC=NC=C2 |
| C9H12N2 | |
| MFCD00006415 | |
| 4-pyrrolidinopyridine, 4-pyrrolidin-1-yl pyridine, 4-1-pyrrolidinyl pyridine, pyridine, 4-1-pyrrolidinyl, 4-pyrrolidin-1-yl-pyridine, 4-pyrrolidinylpyridine, 1-4-pyridyl pyrrolidine, zlchem 148, 4-pyrrolidinepyridine, 4-pyrrollidinopyridine | |
| 4-pyrrolidin-1-ylpyridine |
Specifications
| 2456-81-7 | |
| White-Yellow | |
| C9H12N2 | |
| 5 g | |
| 4-pyrrolidinopyridine, 4-pyrrolidin-1-yl pyridine, 4-1-pyrrolidinyl pyridine, pyridine, 4-1-pyrrolidinyl, 4-pyrrolidin-1-yl-pyridine, 4-pyrrolidinylpyridine, 1-4-pyridyl pyrrolidine, zlchem 148, 4-pyrrolidinepyridine, 4-pyrrollidinopyridine | |
| C1CCN(C1)C2=CC=NC=C2 | |
| 148.209 | |
| 148.21 | |
| Crystalline Powder |
| 58°C | |
| 170°C | |
| MFCD00006415 | |
| 2923 | |
| RGUKYNXWOWSRET-UHFFFAOYSA-N | |
| 4-pyrrolidin-1-ylpyridine | |
| 75567 | |
| ≥98.0% (T) | |
| 4-Pyrrolidinopyridine |
Safety and Handling
TSCA : No