accessibility menu, dialog, popup

UserName

NHN

CAS RN 2456-81-7

IUPAC Name: 4-(pyrrolidin-1-yl)pyridin-1-ium
Synonyms: 4-(pyrrolidin-1-yl)pyridin-1-ium
Molecular Weight (g/mol): 149.22
Molecular Formula: C9H13N2
InChi Key: RGUKYNXWOWSRET-UHFFFAOYSA-O
SMILES: C1CCN(C1)C1=CC=[NH+]C=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)