missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrrolidinopyridine 98.0+%, TCI America™

Catalog No. P09395G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09395G 5 g
P093925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P09395G Supplier TCI America Supplier No. P09395G

Chemical Identifiers

CAS 2456-81-7
Molecular Formula C9H12N2
Molecular Weight (g/mol) 148.209
MDL Number MFCD00006415
InChI Key RGUKYNXWOWSRET-UHFFFAOYSA-N
Synonym 4-pyrrolidinopyridine, 4-pyrrolidin-1-yl pyridine, 4-1-pyrrolidinyl pyridine, pyridine, 4-1-pyrrolidinyl, 4-pyrrolidin-1-yl-pyridine, 4-pyrrolidinylpyridine, 1-4-pyridyl pyrrolidine, zlchem 148, 4-pyrrolidinepyridine, 4-pyrrollidinopyridine
PubChem CID 75567
IUPAC Name 4-pyrrolidin-1-ylpyridine
SMILES C1CCN(C1)C2=CC=NC=C2

Specifications

Melting Point 58°C
Color White-Yellow
Boiling Point 170°C
Quantity 5 g
UN Number 2923
Formula Weight 148.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Pyrrolidinopyridine