missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrrolidinopyridine, 98%

Catalog No. p-189566
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC200010050 5 g Glass bottle
1 options

Catalog No. AC200010050

Supplier: Thermo Scientific Chemicals 200010050

Only null left

CAS: 2456-81-7 | C9H12N2 | 148.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2456-81-7
Molecular Formula C9H12N2
Molecular Weight (g/mol) 148.21
MDL Number MFCD00006415
InChI Key RGUKYNXWOWSRET-UHFFFAOYSA-N
Synonym 4-pyrrolidinopyridine, 4-pyrrolidin-1-yl pyridine, 4-1-pyrrolidinyl pyridine, pyridine, 4-1-pyrrolidinyl, 4-pyrrolidin-1-yl-pyridine, 4-pyrrolidinylpyridine, 1-4-pyridyl pyrrolidine, zlchem 148, 4-pyrrolidinepyridine, 4-pyrrollidinopyridine
PubChem CID 75567
IUPAC Name 4-pyrrolidin-1-ylpyridine
SMILES C1CCN(C1)C2=CC=NC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 54°C to 58°C
Color Brown to White
Boiling Point 171°C to 173°C (27.0 mbar)
Flash Point 142°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Fieser 04,416; 09,400; 10,336; 13,262; 01,1049; 02,377; 03,262; 04,443; 05,597; 06,530; 07,329; 08,449; 09,420; 10,357; 11,477; 12,441; 13,21; 14,99; 15,290; 16,304; 17,189
Solubility Information Solubility in water: 3g/L (20°C). Other solubilities: soluble in acetic acid,soluble in methanol,benzene,ethyl acetate,,chloroform,acetone and dichloroethane,insoluble in hexane and cyclohexane
Formula Weight 148.21
Percent Purity 98%
Physical Form Crystalline Mass, Crystals or Chunks
Chemical Name or Material 4-Pyrrolidinopyridine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 219-534-2

RUO – Research Use Only