missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrimidinemethanol, 98%

Catalog No. AAH31754MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH31754MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH31754MD Supplier Thermo Scientific Chemicals Supplier No. H31754MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 33581-98-5 | C5H6N2O | 110.116 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33581-98-5
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
MDL Number MFCD03789625
InChI Key OEYVFRVNVPKHQQ-UHFFFAOYSA-N
Synonym 4-pyrimidinemethanol, 4-hydroxymethyl pyrimidine, pyrimidin-4-yl methanol, pyrimidin-4-yl-methanol, 4-pyrimidinylmethanol, pyrimidin-4-ylmethan-1-ol, 4-hydroxymethyl-1,3-diazine, pubchem9756, pyrimidine-4-methanol, 4-pyrimidylmethyl alcohol
PubChem CID 12933341
IUPAC Name pyrimidin-4-ylmethanol
SMILES C1=CN=CN=C1CO

Specifications

Melting Point 75°C to 78°C
Quantity 250 mg
Formula Weight 110.12
Percent Purity 98%
Chemical Name or Material 4-Pyrimidinemethanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only