CAS RN 33581-98-5
IUPAC Name:
(pyrimidin-4-yl)methanol
Synonyms:
pyrimidin-4-ylmethanol
Molecular Weight (g/mol):
110.12
Molecular Formula:
C5H6N2O
InChi Key:
OEYVFRVNVPKHQQ-UHFFFAOYSA-N
SMILES:
OCC1=NC=NC=C1
1
–
1
of
1
results
1
Promotions Available
4-Pyrimidinemethanol, 98%
4-Pyrimidinemethanol, 98%
CAS: 33581-98-5 Molecular Formula: C5H6N2O Molecular Weight (g/mol): 110.116 MDL Number: MFCD03789625 InChI Key: OEYVFRVNVPKHQQ-UHFFFAOYSA-N Synonym: 4-pyrimidinemethanol,4-hydroxymethyl pyrimidine,pyrimidin-4-yl methanol,pyrimidin-4-yl-methanol,4-pyrimidinylmethanol,pyrimidin-4-ylmethan-1-ol,4-hydroxymethyl-1,3-diazine,pubchem9756,pyrimidine-4-methanol,4-pyrimidylmethyl alcohol PubChem CID: 12933341 IUPAC Name: pyrimidin-4-ylmethanol SMILES: C1=CN=CN=C1CO