missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrimidin-2-ylbenzaldehyde, 95%, Thermo Scientific™

Chemical Identifiers

CAS 77232-38-3
Molecular Formula C11H8N2O
Molecular Weight (g/mol) 184.20
MDL Number MFCD08056282
InChI Key JUYLMPWKVWDXBE-UHFFFAOYSA-N
Synonym 4-pyrimidin-2-yl benzaldehyde, 4-2-pyrimidinyl benzaldehyde, 2-4-formylphenyl pyrimidine, 4-pyrimidin-2-yl-benzaldehyde, 4-2-pyrimidinyl-benzaldehyde, benzaldehyde,4-2-pyrimidinyl, benzaldehyde, 4-2-pyrimidinyl
PubChem CID 12703722
IUPAC Name 4-pyrimidin-2-ylbenzaldehyde
SMILES O=CC1=CC=C(C=C1)C1=NC=CC=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC56204DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC56204DE
View Documents Promotion Details
N/A 5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

77232-38-3
184.20
JUYLMPWKVWDXBE-UHFFFAOYSA-N
12703722
O=CC1=CC=C(C=C1)C1=NC=CC=N1
C11H8N2O
MFCD08056282
4-pyrimidin-2-yl benzaldehyde, 4-2-pyrimidinyl benzaldehyde, 2-4-formylphenyl pyrimidine, 4-pyrimidin-2-yl-benzaldehyde, 4-2-pyrimidinyl-benzaldehyde, benzaldehyde,4-2-pyrimidinyl, benzaldehyde, 4-2-pyrimidinyl
4-pyrimidin-2-ylbenzaldehyde

Specifications

77232-38-3
100.0
C11H8N2O
4-pyrimidin-2-yl benzaldehyde, 4-2-pyrimidinyl benzaldehyde, 2-4-formylphenyl pyrimidine, 4-pyrimidin-2-yl-benzaldehyde, 4-2-pyrimidinyl-benzaldehyde, benzaldehyde,4-2-pyrimidinyl, benzaldehyde, 4-2-pyrimidinyl
O=CC1=CC=C(C=C1)C1=NC=CC=N1
184.20
184.2
4-Pyrimidin-2-ylbenzaldehyde
94.5
95%
MFCD08056282
JUYLMPWKVWDXBE-UHFFFAOYSA-N
4-pyrimidin-2-ylbenzaldehyde
12703722
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents