accessibility menu, dialog, popup

UserName

NNO

CAS RN 77232-38-3

IUPAC Name: 4-(pyrimidin-2-yl)benzaldehyde
Synonyms: 4-(pyrimidin-2-yl)benzaldehyde
Molecular Weight (g/mol): 184.2
Molecular Formula: C11H8N2O
InChi Key: JUYLMPWKVWDXBE-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)C1=NC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)