Learn More
4-Pyridinemethanol, 99%
CAS: 586-95-8 | C6H7NO | 109.13 g/mol
Supplier: Thermo Scientific Chemicals A1469822
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Pyridinemethanol | |
| 53°C to 55°C | |
| 148°C (298°F) | |
| MFCD00006442 | |
| 107850 | |
| 4-pyridinemethanol, 4-pyridylcarbinol, 4-hydroxymethyl pyridine, pyridine-4-methanol, 4-pyridylmethanol, 4-pyridinylmethanol, 4-pyridinecarbinol, 4-picolyl alcohol, 4-hydroxymethylpyridine, pyridin-4-yl methanol | |
| OCC1=CC=NC=C1 | |
| 109.13 | |
| 109.13 |
| 586-95-8 | |
| 141°C (12 mmHg) | |
| C6H7NO | |
| 100 g | |
| Hygroscopic | |
| PTMBWNZJOQBTBK-UHFFFAOYSA-N | |
| pyridin-4-ylmethanol | |
| 11472 | |
| 99% |
Chemical Identifiers
| 586-95-8 | |
| 109.13 | |
| PTMBWNZJOQBTBK-UHFFFAOYSA-N | |
| 11472 |
| C6H7NO | |
| MFCD00006442 | |
| 4-pyridinemethanol, 4-pyridylcarbinol, 4-hydroxymethyl pyridine, pyridine-4-methanol, 4-pyridylmethanol, 4-pyridinylmethanol, 4-pyridinecarbinol, 4-picolyl alcohol, 4-hydroxymethylpyridine, pyridin-4-yl methanol | |
| OCC1=CC=NC=C1 |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 209-590-6
RTECSNumber : UT4691000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only