missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pyridinemethanol 97.0+%, TCI America™
$179.05 - $179.05
Chemical Identifiers
| CAS | 586-95-8 |
|---|---|
| Molecular Formula | C6H7NO |
| Molecular Weight (g/mol) | 109.13 |
| MDL Number | MFCD00006442 |
| InChI Key | PTMBWNZJOQBTBK-UHFFFAOYSA-N |
| Synonym | 4-pyridinemethanol, 4-pyridylcarbinol, 4-hydroxymethyl pyridine, pyridine-4-methanol, 4-pyridylmethanol, 4-pyridinylmethanol, 4-pyridinecarbinol, 4-picolyl alcohol, 4-hydroxymethylpyridine, pyridin-4-yl methanol |
| PubChem CID | 11472 |
| IUPAC Name | (pyridin-4-yl)methanol |
| SMILES | OCC1=CC=NC=C1 |
Chemical Identifiers
| 586-95-8 | |
| 109.13 | |
| PTMBWNZJOQBTBK-UHFFFAOYSA-N | |
| 11472 | |
| OCC1=CC=NC=C1 |
| C6H7NO | |
| MFCD00006442 | |
| 4-pyridinemethanol, 4-pyridylcarbinol, 4-hydroxymethyl pyridine, pyridine-4-methanol, 4-pyridylmethanol, 4-pyridinylmethanol, 4-pyridinecarbinol, 4-picolyl alcohol, 4-hydroxymethylpyridine, pyridin-4-yl methanol | |
| (pyridin-4-yl)methanol |
Specifications
| 586-95-8 | |
| 150°C | |
| MFCD00006442 | |
| 4-pyridinemethanol, 4-pyridylcarbinol, 4-hydroxymethyl pyridine, pyridine-4-methanol, 4-pyridylmethanol, 4-pyridinylmethanol, 4-pyridinecarbinol, 4-picolyl alcohol, 4-hydroxymethylpyridine, pyridin-4-yl methanol | |
| OCC1=CC=NC=C1 | |
| 109.13 | |
| 109.13 | |
| Crystalline Powder |
| 55°C | |
| C6H7NO | |
| 25 g | |
| PTMBWNZJOQBTBK-UHFFFAOYSA-N | |
| (pyridin-4-yl)methanol | |
| 11472 | |
| ≥97.0% (GC) | |
| 4-Pyridinemethanol |
Safety and Handling
EINECSNumber : (9)-1112
RTECSNumber : UT4691000
TSCA : Yes