missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pyridineethanol Hydrochloride 98.0+%, TCI America™
Supplier: TCI America P129325G
Specifications
| 4-Pyridineethanol Hydrochloride | |
| 151°C | |
| C7H10ClNO | |
| 25 g | |
| SUDVBWMNUVBMHV-UHFFFAOYSA-N | |
| 2-(pyridin-4-yl)ethan-1-ol hydrochloride | |
| 23090609 | |
| ≥98.0% (HPLC,T) |
| 383177-54-6 | |
| White | |
| MFCD01321183 | |
| 4-(2-Hydroxyethyl)pyridine Hydrochloride, 2-(4-Pyridyl)ethanol Hydrochloride | |
| Cl.OCCC1=CC=NC=C1 | |
| 159.61 | |
| 159.61 | |
| Crystalline Powder |
Chemical Identifiers
| 383177-54-6 | |
| 159.61 | |
| SUDVBWMNUVBMHV-UHFFFAOYSA-N | |
| 23090609 | |
| Cl.OCCC1=CC=NC=C1 |
| C7H10ClNO | |
| MFCD01321183 | |
| 4-(2-Hydroxyethyl)pyridine Hydrochloride, 2-(4-Pyridyl)ethanol Hydrochloride | |
| 2-(pyridin-4-yl)ethan-1-ol hydrochloride |
Safety and Handling
TSCA : No