missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridineacetamide, 98%

Catalog No. AAB2473406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2473406 5 g
AAB2473403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2473406 Supplier Thermo Scientific Chemicals Supplier No. B2473406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39640-62-5 | C7H8N2O | 136.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39640-62-5
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD02685124
InChI Key YEALAJQPOVKTOH-UHFFFAOYSA-N
Synonym 4-pyridineacetamide, 2-pyridin-4-yl acetamide, pyridine-4-acetamide, acmc-20ajsz, 4-pyridyl acetamide, 4-pyridylacetic acid amide, 2-pyridine-4-yl-acetamide
PubChem CID 227128
IUPAC Name 2-pyridin-4-ylacetamide
SMILES C1=CN=CC=C1CC(=O)N

Specifications

Melting Point 144°C to 147°C
Quantity 5 g
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material 4-Pyridineacetamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only