CAS RN 39640-62-5
IUPAC Name:
2-(pyridin-4-yl)acetamide
Synonyms:
4-pyridineacetamide
Molecular Weight (g/mol):
136.15
Molecular Formula:
C7H8N2O
InChi Key:
YEALAJQPOVKTOH-UHFFFAOYSA-N
SMILES:
NC(=O)CC1=CC=NC=C1
1
–
1
of
1
results
1
Promotions Available
4-Pyridineacetamide, 98%
4-Pyridineacetamide, 98%
CAS: 39640-62-5 Molecular Formula: C7H8N2O Molecular Weight (g/mol): 136.154 MDL Number: MFCD02685124 InChI Key: YEALAJQPOVKTOH-UHFFFAOYSA-N Synonym: 4-pyridineacetamide,2-pyridin-4-yl acetamide,pyridine-4-acetamide,acmc-20ajsz,4-pyridyl acetamide,4-pyridylacetic acid amide,2-pyridine-4-yl-acetamide PubChem CID: 227128 IUPAC Name: 2-pyridin-4-ylacetamide SMILES: C1=CN=CC=C1CC(=O)N