missing translation for 'onlineSavingsMsg'
Learn More

4-Propylpyridine 97.0+%, TCI America™

Catalog No. P06695ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06695ML 5 mL
P066925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P06695ML Supplier TCI America Supplier No. P06695ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1122-81-2
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00014639
InChI Key JAWZAONCXMJLFT-UHFFFAOYSA-N
Synonym 4-n-propylpyridine, pyridine, 4-propyl, 4-n-propylpylpyridine, 4-n-propyl-pyridine, 4-propyl-pyridin, 4-propyl pyridine, pubchem15021, 1-4-pyridyl propane, 1-pyridin-4-yl propane, 4-n-propylpyridine 97+%
PubChem CID 70738
IUPAC Name 4-propylpyridine
SMILES CCCC1=CC=NC=C1

Specifications

Color Yellow
Boiling Point 190°C
Quantity 5 mL
Formula Weight 121.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Propylpyridine
EINECSNumber (9)-1843
TSCA Yes