accessibility menu, dialog, popup

UserName

CAS RN 1122-81-2

IUPAC Name: 4-propylpyridine
Synonyms: 4-propylpyridine
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: JAWZAONCXMJLFT-UHFFFAOYSA-N
SMILES: CCCC1=CC=NC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results