missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Propylphenol 99.0+%, TCI America™
$94.33 - $227.16
Chemical Identifiers
| CAS | 645-56-7 |
|---|---|
| Molecular Formula | C9H12O |
| Molecular Weight (g/mol) | 136.194 |
| MDL Number | MFCD00002395 |
| InChI Key | KLSLBUSXWBJMEC-UHFFFAOYSA-N |
| Synonym | 4-n-propylphenol, p-propylphenol, phenol, 4-propyl, dihydrochavicol, phenol, p-propyl, 1-hydroxy-4-n-propylbenzene, p-n-propylphenol, 4-propyl-phenol, p-hydroxypropylbenzene, hydroxypropylbenzene, p |
| PubChem CID | 12580 |
| ChEBI | CHEBI:34434 |
| IUPAC Name | 4-propylphenol |
| SMILES | CCCC1=CC=C(C=C1)O |
Chemical Identifiers
| 645-56-7 | |
| 136.194 | |
| KLSLBUSXWBJMEC-UHFFFAOYSA-N | |
| 12580 | |
| 4-propylphenol |
| C9H12O | |
| MFCD00002395 | |
| 4-n-propylphenol, p-propylphenol, phenol, 4-propyl, dihydrochavicol, phenol, p-propyl, 1-hydroxy-4-n-propylbenzene, p-n-propylphenol, 4-propyl-phenol, p-hydroxypropylbenzene, hydroxypropylbenzene, p | |
| CHEBI:34434 | |
| CCCC1=CC=C(C=C1)O |
Specifications
| 645-56-7 | |
| 232°C | |
| MFCD00002395 | |
| 2430 | |
| KLSLBUSXWBJMEC-UHFFFAOYSA-N | |
| 4-propylphenol | |
| 12580 | |
| 136.19 | |
| Crystalline Lumps |
| White-Yellow | |
| C9H12O | |
| 5 g | |
| 4-n-propylphenol, p-propylphenol, phenol, 4-propyl, dihydrochavicol, phenol, p-propyl, 1-hydroxy-4-n-propylbenzene, p-n-propylphenol, 4-propyl-phenol, p-hydroxypropylbenzene, hydroxypropylbenzene, p | |
| CCCC1=CC=C(C=C1)O | |
| 136.194 | |
| CHEBI:34434 | |
| ≥99.0% (GC) | |
| 4-Propylphenol |
Safety and Handling
EINECSNumber : (3)-0503&(4)-0057
RTECSNumber : SM8610000
TSCA : Yes