missing translation for 'onlineSavingsMsg'
Learn More

4-Propylphenol 99.0+%, TCI America™

Catalog No. P06985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06985G 5 g
P069825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P06985G Supplier TCI America Supplier No. P06985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 645-56-7
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002395
InChI Key KLSLBUSXWBJMEC-UHFFFAOYSA-N
Synonym 4-n-propylphenol, p-propylphenol, phenol, 4-propyl, dihydrochavicol, phenol, p-propyl, 1-hydroxy-4-n-propylbenzene, p-n-propylphenol, 4-propyl-phenol, p-hydroxypropylbenzene, hydroxypropylbenzene, p
PubChem CID 12580
ChEBI CHEBI:34434
IUPAC Name 4-propylphenol
SMILES CCCC1=CC=C(C=C1)O

Specifications

Color White-Yellow
Boiling Point 232°C
Quantity 5 g
UN Number 2430
Formula Weight 136.19
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Propylphenol
EINECSNumber (3)-0503&(4)-0057
RTECSNumber SM8610000
TSCA Yes