missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Propylcyclohexanone 98.0+%, TCI America™
$36.57 - $90.48
Chemical Identifiers
| CAS | 40649-36-3 |
|---|---|
| Molecular Formula | C9H16O |
| Molecular Weight (g/mol) | 140.226 |
| MDL Number | MFCD00058690 |
| InChI Key | NQEDLIZOPMNZMC-UHFFFAOYSA-N |
| Synonym | 4-propylcyclohexanone, 4-n-propylcyclohexanone, cyclohexanone, 4-propyl, 4-propyl-1-cyclohexanone, 4-propylcyclohexanon, 4-propyl-cyclohexanone, pubchem14023, acmc-1arvj, 4beta-propylcyclohexanone, 4-n-propyl-cyclohexanone |
| PubChem CID | 142482 |
| IUPAC Name | 4-propylcyclohexan-1-one |
| SMILES | CCCC1CCC(=O)CC1 |
Chemical Identifiers
| 40649-36-3 | |
| 140.226 | |
| NQEDLIZOPMNZMC-UHFFFAOYSA-N | |
| 142482 | |
| CCCC1CCC(=O)CC1 |
| C9H16O | |
| MFCD00058690 | |
| 4-propylcyclohexanone, 4-n-propylcyclohexanone, cyclohexanone, 4-propyl, 4-propyl-1-cyclohexanone, 4-propylcyclohexanon, 4-propyl-cyclohexanone, pubchem14023, acmc-1arvj, 4beta-propylcyclohexanone, 4-n-propyl-cyclohexanone | |
| 4-propylcyclohexan-1-one |
Specifications
| 40649-36-3 | |
| 98°C | |
| MFCD00058690 | |
| 4-propylcyclohexanone, 4-n-propylcyclohexanone, cyclohexanone, 4-propyl, 4-propyl-1-cyclohexanone, 4-propylcyclohexanon, 4-propyl-cyclohexanone, pubchem14023, acmc-1arvj, 4beta-propylcyclohexanone, 4-n-propyl-cyclohexanone | |
| CCCC1CCC(=O)CC1 | |
| 140.226 | |
| 140.23 | |
| Liquid |
| Colorless | |
| C9H16O | |
| 5 g | |
| NQEDLIZOPMNZMC-UHFFFAOYSA-N | |
| 4-propylcyclohexan-1-one | |
| 142482 | |
| ≥98.0% (GC) | |
| 4-Propylcyclohexanone |
Safety and Handling
EINECSNumber : (3)-2411
TSCA : No