missing translation for 'onlineSavingsMsg'
Learn More

4-Propylcyclohexanone 98.0+%, TCI America™

Catalog No. p184925g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
P184925G 25 g
P18495G 5 g
2 options

Catalog No. P184925G

Supplier: TCI America P184925G

Only null left

Chemical Identifiers

CAS 40649-36-3
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.226
MDL Number MFCD00058690
InChI Key NQEDLIZOPMNZMC-UHFFFAOYSA-N
Synonym 4-propylcyclohexanone, 4-n-propylcyclohexanone, cyclohexanone, 4-propyl, 4-propyl-1-cyclohexanone, 4-propylcyclohexanon, 4-propyl-cyclohexanone, pubchem14023, acmc-1arvj, 4beta-propylcyclohexanone, 4-n-propyl-cyclohexanone
PubChem CID 142482
IUPAC Name 4-propylcyclohexan-1-one
SMILES CCCC1CCC(=O)CC1

Specifications

Color Colorless
Boiling Point 98°C
Quantity 25 g
Formula Weight 140.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Propylcyclohexanone
EINECSNumber (3)-2411
TSCA No