missing translation for 'onlineSavingsMsg'
Learn More

4-Propylcyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. p-7135193
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16565G 5 g
P165625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16565G Supplier TCI America Supplier No. P16565G
Only null left

Chemical Identifiers

CAS 70928-91-5
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
InChI Key QCNUKEGGHOLBES-UHFFFAOYSA-N
PubChem CID 1490324
IUPAC Name 4-propylcyclohexane-1-carboxylic acid
SMILES CCCC1CCC(CC1)C(=O)O

Specifications

Color Colorless
Quantity 5 g
Formula Weight 170.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Propylcyclohexanecarboxylic Acid (cis- and trans- mixture)
EINECSNumber (3)-3723
RTECSNumber GU8648000
TSCA No