accessibility menu, dialog, popup

UserName

CAS RN 70928-91-5

CH3OHO

CAS RN 70928-91-5

IUPAC Name: 4-propylcyclohexane-1-carboxylic acid
Synonyms: 4-propylcyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: QCNUKEGGHOLBES-UHFFFAOYSA-N
SMILES: CCCC1CCC(CC1)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results