missing translation for 'onlineSavingsMsg'
Learn More

4-Propoxyphthalonitrile 95.0+%, TCI America™

Catalog No. P10701G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10701G 1 g
P10705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P10701G Supplier TCI America Supplier No. P10701G

Chemical Identifiers

CAS 106144-18-7
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.214
MDL Number MFCD00059102
InChI Key RDLZRCSOLOTEPK-UHFFFAOYSA-N
Synonym 1,2-Dicyano-4-propoxybenzene
PubChem CID 588896
IUPAC Name 4-propoxybenzene-1,2-dicarbonitrile
SMILES CCCOC1=CC(=C(C=C1)C#N)C#N

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 186.21
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Propoxyphthalonitrile