missing translation for 'onlineSavingsMsg'
Learn More

4-Propoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p19685g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P19685G Supplier TCI America Supplier No. P19685G

Chemical Identifiers

CAS 186497-67-6
Molecular Formula C9H13BO3
Molecular Weight (g/mol) 180.01
MDL Number MFCD03427189
InChI Key SOKFEJKZYHYKSY-UHFFFAOYSA-N
Synonym 4-propoxybenzeneboronic acid, 4-n-propoxyphenylboronic acid, 4-propoxyphenyl boronic acid, 4-propyloxyphenyl boronic acid, 4-n-propoxybenzeneboronic acid, p-propoxyphenylboronic acid, 4-propoxyphenylboronicacid, boronic acid, 4-propoxyphenyl, b-4-propoxyphenyl boronic acid, pubchem9546
PubChem CID 4456490
IUPAC Name (4-propoxyphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)OCCC)(O)O

Specifications

Melting Point 126°C
Color White
Quantity 5 g
Formula Weight 180.01
Physical Form Crystalline Powder
Chemical Name or Material 4-Propoxyphenylboronic Acid (contains varying amounts of Anhydride)