accessibility menu, dialog, popup

UserName

H3COBOHHO

CAS RN 186497-67-6

IUPAC Name: (4-propoxyphenyl)boronic acid
Synonyms: 4-propoxyphenylboronic acid
Molecular Weight (g/mol): 180.01
Molecular Formula: C9H13BO3
InChi Key: SOKFEJKZYHYKSY-UHFFFAOYSA-N
SMILES: CCCOC1=CC=C(C=C1)B(O)O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results