missing translation for 'onlineSavingsMsg'
Learn More

4-Propoxybenzaldehyde 97.0+%, TCI America™

Catalog No. P018225ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P018225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P018225ML Supplier TCI America Supplier No. P018225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5736-85-6
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00014134
InChI Key FGXZWMCBNMMYPL-UHFFFAOYSA-N
Synonym p-propoxybenzaldehyde, benzaldehyde, 4-propoxy, 4-n-propoxybenzaldehyde, propoxybenzaldehyde, benzaldehyde, p-propoxy, p-n-propoxy benzaldehyde, p-n-propoxybenzaldehyde, 4-propoxybenzaldehyde, 4-n-propyloxybenzaldehyde, wln: vhr do3
PubChem CID 79812
IUPAC Name 4-propoxybenzaldehyde
SMILES CCCOC1=CC=C(C=C1)C=O

Specifications

Color Yellow
Boiling Point 112°C
Quantity 25 mL
Formula Weight 164.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Propoxybenzaldehyde
RTECSNumber CU7700000
TSCA Yes