accessibility menu, dialog, popup

UserName

H3COO

CAS RN 5736-85-6

IUPAC Name: 4-propoxybenzaldehyde
Synonyms: benzaldehyde, 4-propoxy-
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: FGXZWMCBNMMYPL-UHFFFAOYSA-N
SMILES: CCCOC1=CC=C(C=O)C=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)