missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Propargyloxy)benzaldehyde 98.0+%, TCI America™
Supplier: TCI America P23391G
Specifications
| 4-(Propargyloxy)benzaldehyde | |
| 82°C | |
| MFCD01088150 | |
| 4-(2-Propynyloxy)benzaldehyde | |
| O=CC1=CC=C(OCC#C)C=C1 | |
| 160.17 | |
| 160.17 | |
| Crystalline Powder |
| 5651-86-5 | |
| C10H8O2 | |
| 1 g | |
| GSSBOYWRKTVVQX-UHFFFAOYSA-N | |
| 4-(prop-2-yn-1-yloxy)benzaldehyde | |
| 1897533 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 5651-86-5 | |
| 160.17 | |
| GSSBOYWRKTVVQX-UHFFFAOYSA-N | |
| 1897533 | |
| O=CC1=CC=C(OCC#C)C=C1 |
| C10H8O2 | |
| MFCD01088150 | |
| 4-(2-Propynyloxy)benzaldehyde | |
| 4-(prop-2-yn-1-yloxy)benzaldehyde |
Safety and Handling
TSCA : No