CAS RN 5651-86-5
IUPAC Name:
4-(prop-2-yn-1-yloxy)benzaldehyde
Synonyms:
4-(prop-2-yn-1-yloxy)benzaldehyde
Molecular Weight (g/mol):
160.17
Molecular Formula:
C10H8O2
InChi Key:
GSSBOYWRKTVVQX-UHFFFAOYSA-N
SMILES:
O=CC1=CC=C(OCC#C)C=C1
1
–
1
of
1
results
1
4-(Propargyloxy)benzaldehyde 98.0+%, TCI America™
CAS: 5651-86-5 Molecular Formula: C10H8O2 Molecular Weight (g/mol): 160.17 MDL Number: MFCD01088150 InChI Key: GSSBOYWRKTVVQX-UHFFFAOYSA-N Synonym: 4-(2-Propynyloxy)benzaldehyde PubChem CID: 1897533 IUPAC Name: 4-(prop-2-yn-1-yloxy)benzaldehyde SMILES: O=CC1=CC=C(OCC#C)C=C1