missing translation for 'onlineSavingsMsg'
Learn More

4-(Propargyloxy)benzaldehyde 98.0+%, TCI America™

Catalog No. P2339200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2339200MG 200 mg
P23391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2339200MG Supplier TCI America Supplier No. P2339200MG
Only null left

Chemical Identifiers

CAS 5651-86-5
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
MDL Number MFCD01088150
InChI Key GSSBOYWRKTVVQX-UHFFFAOYSA-N
Synonym 4-(2-Propynyloxy)benzaldehyde
PubChem CID 1897533
IUPAC Name 4-(prop-2-yn-1-yloxy)benzaldehyde
SMILES O=CC1=CC=C(OCC#C)C=C1

Specifications

Melting Point 82°C
Quantity 200 mg
Formula Weight 160.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Propargyloxy)benzaldehyde