missing translation for 'onlineSavingsMsg'
Learn More

4-Piperidinopiperidine 98.0+%, TCI America™

Catalog No. P16385G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16385G 5 g
P16381G 1 g
P163825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P16385G Supplier TCI America Supplier No. P16385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4897-50-1
Molecular Formula C10H20N2
Molecular Weight (g/mol) 168.284
InChI Key QDVBKXJMLILLLB-UHFFFAOYSA-N
Synonym 4-piperidinopiperidine, 1,4'-bipiperidine, 4-piperidino-piperidine, 1,4'-bipiperidyl, 4-1-piperidinyl piperidine, 1,4' bipiperidinyl, 4-piperidin-1-yl piperidine, 1,4-bipiperidine, 1-piperidin-4-yl piperidine, 4-piperidin-1-ylpiperidine
PubChem CID 78607
ChEBI CHEBI:40117
IUPAC Name 1-piperidin-4-ylpiperidine
SMILES C1CCN(CC1)C2CCNCC2

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 5 g
Formula Weight 168.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Piperidinopiperidine