missing translation for 'onlineSavingsMsg'
Learn More

4-Picolylamine 98.0+%, TCI America™

Catalog No. P077925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P077925G 25 g
P0779100G 100 g
P0779500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P077925G Supplier TCI America Supplier No. P077925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3731-53-1
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006449
InChI Key TXQWFIVRZNOPCK-UHFFFAOYSA-N
Synonym 4-pyridinemethaneamine, 4-aminomethyl pyridine, 4-pyridinemethanamine, 4-pyridylmethylamine, 4-picolylamine, 4-aminomethylpyridine, 4-picolinylamine, 4-pyridinemethylamine, 1-pyridin-4-ylmethanamine, 4-picolinamine
PubChem CID 77317
IUPAC Name pyridin-4-ylmethanamine
SMILES C1=CN=CC=C1CN

Specifications

Melting Point -8°C
Boiling Point 230°C
Quantity 25 g
UN Number 2735
Formula Weight 108.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Picolylamine
EINECSNumber (5)-3701
TSCA Yes