accessibility menu, dialog, popup

UserName

CAS RN 3731-53-1

IUPAC Name: 1-(pyridin-4-yl)methanamine
Synonyms: 4-pyridinemethaneamine
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: TXQWFIVRZNOPCK-UHFFFAOYSA-N
SMILES: NCC1=CC=NC=C1
Boiling Point 
  • (3)
  • (5)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (3)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (5)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)