missing translation for 'onlineSavingsMsg'
Learn More

4-Picolyl chloride hydrochloride, 97%

Catalog No. p-184756
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC131200100 10 g Glass bottle
AC131201000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC131200100 Supplier Thermo Scientific Chemicals Supplier No. 131200100

CAS: 1822-51-1 | C6H6ClN·HCl | 164.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1822-51-1
Molecular Formula C6H6ClN·HCl
Molecular Weight (g/mol) 164.04
MDL Number MFCD00012826
InChI Key ZDHKVKPZQKYREU-UHFFFAOYSA-N
Synonym 4-chloromethyl pyridine hydrochloride, 4-picolyl chloride hydrochloride, 4-chloromethyl pyridinium chloride, 4-chloromethylpyridine hydrochloride, 4-chloromethyl-pyridine hydrochloride, ccris 1728, isonicotinyl chloride hydrochloride, pyridine, 4-chloromethyl-, hydrochloride, 4-chloromethyl pyridine hcl, 4-chloromethyl pyridine, chloride
PubChem CID 74570
IUPAC Name 4-(chloromethyl)pyridine;hydrochloride
SMILES C1=CN=CC=C1CCl.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 160°C to 163°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 10 g
Fieser 04,391
Solubility Information Solubility in water: 750g/L (20°C)
Formula Weight 164.04
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Picolyl chloride hydrochloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-350-7

RUO – Research Use Only