missing translation for 'onlineSavingsMsg'
Learn More

4-Picoline N-oxide, 98%

Catalog No. aaa1733014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1733014 25 g
AAA1733022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1733014 Supplier Thermo Scientific Chemicals Supplier No. A1733014
Only null left

CAS: 1003-67-4 | C6H7NO | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 1003-67-4
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006210
InChI Key IWYYIZOHWPCALJ-UHFFFAOYSA-N
Synonym 4-picoline-n-oxide, 4-picoline n-oxide, 4-methylpyridine n-oxide, 4-methylpyridine 1-oxide, pyridine, 4-methyl-, 1-oxide, 4-picoline 1-oxide, 4-picoline, 1-oxide, gamma-picoline n-oxide, gamma-picoline 1-oxide, 4-methylpyridin-1-ium-1-olate
PubChem CID 13857
SMILES CC1=CC=[N+]([O-])C=C1

Specifications

Melting Point 182°C to 185°C
Boiling Point 152°C (11 mmHg)
Flash Point 149°C (300°F)
Quantity 25 g
Beilstein 106329
Sensitivity Hygroscopic
IUPAC Name 4-methyl-1-oxidopyridin-1-ium
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 4-Picoline N-oxide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-712-3
RTECSNumber UT5800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only