accessibility menu, dialog, popup

UserName

CH3NO

CAS RN 1003-67-4

IUPAC Name: 4-methylpyridin-1-ium-1-olate
Synonyms: 4-methylpyridine N-oxide
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: IWYYIZOHWPCALJ-UHFFFAOYSA-N
SMILES: CC1=CC=[N+]([O-])C=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)