missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylurazole 98.0+%, TCI America™

Catalog No. p-7134983
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11605G 5 g
P116025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11605G Supplier TCI America Supplier No. P11605G
Only null left

Chemical Identifiers

CAS 15988-11-1
Molecular Formula C8H7N3O2
Molecular Weight (g/mol) 177.163
MDL Number MFCD00005226
InChI Key GOSUFRDROXZXLN-UHFFFAOYSA-N
Synonym 4-phenylurazole, urazole, 4-phenyl, 1,2,4-triazolidine-3,5-dione, 4-phenyl, bicarbamimide, n-phenyl, 4-phenyl urazole, 4-phenylurazol, 4-phenyl-urazole, acmc-1c52y, ksc297q5n
PubChem CID 85229
IUPAC Name 4-phenyl-1,2,4-triazolidine-3,5-dione
SMILES C1=CC=C(C=C1)N2C(=O)NNC2=O

Specifications

Melting Point 209°C
Color White
Quantity 5 g
Formula Weight 177.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylurazole