accessibility menu, dialog, popup

UserName

ONHNHON

CAS RN 15988-11-1

IUPAC Name: 4-phenyl-1,2,4-triazolidine-3,5-dione
Synonyms: 4-phenyl-1,2,4-triazolidine-3,5-dione
Molecular Weight (g/mol): 177.16
Molecular Formula: C8H7N3O2
InChi Key: GOSUFRDROXZXLN-UHFFFAOYSA-N
SMILES: O=C1NNC(=O)N1C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)