missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylurazole, 98+%

Catalog No. AAL0025506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0025506 5 g
AAL0025514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0025506 Supplier Thermo Scientific Chemicals Supplier No. L0025506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 15988-11-1 | C8H7N3O2 | 177.163 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15988-11-1
Molecular Formula C8H7N3O2
Molecular Weight (g/mol) 177.163
MDL Number MFCD00005226
InChI Key GOSUFRDROXZXLN-UHFFFAOYSA-N
Synonym 4-phenylurazole, urazole, 4-phenyl, 1,2,4-triazolidine-3,5-dione, 4-phenyl, bicarbamimide, n-phenyl, 4-phenyl urazole, 4-phenylurazol, 4-phenyl-urazole, acmc-1c52y, ksc297q5n
PubChem CID 85229
IUPAC Name 4-phenyl-1,2,4-triazolidine-3,5-dione
SMILES C1=CC=C(C=C1)N2C(=O)NNC2=O

Specifications

Melting Point 206°C to 210°C
Quantity 5 g
Beilstein 163361
Formula Weight 177.16
Percent Purity ≥98%
Chemical Name or Material 4-Phenylurazole
EINECSNumber 240-127-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only