missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylsemicarbazide 98.0+%, TCI America™

Catalog No. P02285G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P02285G 5 g
P022825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P02285G Supplier TCI America Supplier No. P02285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 537-47-3
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.169
MDL Number MFCD00007590
InChI Key MOCKWYUCPREFCZ-UHFFFAOYSA-N
Synonym 4-phenylsemicarbazide, n-phenylhydrazinecarboxamide, 3-amino-1-phenylurea, hydrazinecarboxamide, n-phenyl, anilinoformylhydrazine, unii-bpu96sx57a, semicarbazide, 4-phenyl, bpu96sx57a, hydrazinecarboxanilide, anilinocarbonylhydrazine
PubChem CID 10837
IUPAC Name 1-amino-3-phenylurea
SMILES C1=CC=C(C=C1)NC(=O)NN

Specifications

Melting Point 125°C
Color White
Quantity 5 g
Formula Weight 151.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylsemicarbazide
EINECSNumber (3)-4034
TSCA No