missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylphenol 99.0+%, TCI America™

Catalog No. p-7134483
Change view
Click to view available options
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
P020125G 25 g
P0201100G 100 g
P0201500G 500 g
3 options

Catalog No. P020125G

Supplier: TCI America P020125G

Only null left

Chemical Identifiers

CAS 92-69-3
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.211
MDL Number MFCD00002347
InChI Key YXVFYQXJAXKLAK-UHFFFAOYSA-N
Synonym 4-hydroxybiphenyl, 1,1'-biphenyl-4-ol, biphenyl-4-ol, p-phenylphenol, 4-biphenylol, p-hydroxybiphenyl, 4-hydroxydiphenyl, p-hydroxydiphenyl, p-biphenylol, 4-diphenylol
PubChem CID 7103
ChEBI CHEBI:34422
IUPAC Name 4-phenylphenol
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)O

Specifications

Melting Point 165°C
Color White
Boiling Point 321°C
Quantity 25 g
UN Number 3077
Formula Weight 170.21
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylphenol
EINECSNumber (4)-0019
RTECSNumber DV5850000
TSCA Yes