missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenylmorpholine 98.0+%, TCI America™
Supplier: TCI America P0196100G
Specifications
| 4-Phenylmorpholine | |
| 54°C | |
| C10H13NO | |
| 100 g | |
| n-phenylmorpholine, phenylmorpholine, morpholine, 4-phenyl, morpholinobenzene, 4-phenyl-morpholine, phenyl morpholine, 4-phenylmorpholin, unii-h4oc7fb1ex, h4oc7fb1ex, n-phenyl morpholine | |
| C1CN(CCO1)C1=CC=CC=C1 | |
| 163.22 | |
| 163.22 | |
| Crystalline Powder |
| 92-53-5 | |
| 270°C | |
| MFCD00006166,MFCD31699968 | |
| 2811 | |
| FHQRDEDZJIFJAL-UHFFFAOYSA-N | |
| 4-phenylmorpholine | |
| 62339 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 92-53-5 | |
| 163.22 | |
| FHQRDEDZJIFJAL-UHFFFAOYSA-N | |
| 62339 | |
| C1CN(CCO1)C1=CC=CC=C1 |
| C10H13NO | |
| MFCD00006166,MFCD31699968 | |
| n-phenylmorpholine, phenylmorpholine, morpholine, 4-phenyl, morpholinobenzene, 4-phenyl-morpholine, phenyl morpholine, 4-phenylmorpholin, unii-h4oc7fb1ex, h4oc7fb1ex, n-phenyl morpholine | |
| 4-phenylmorpholine |
Safety and Handling
RTECSNumber : QE8575000
TSCA : Yes