missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylmorpholine 98.0+%, TCI America™

Catalog No. p-7134479
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P019625G 25 g
P0196100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P019625G Supplier TCI America Supplier No. P019625G
Only null left

Chemical Identifiers

CAS 92-53-5
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00006166,MFCD31699968
InChI Key FHQRDEDZJIFJAL-UHFFFAOYSA-N
Synonym n-phenylmorpholine, phenylmorpholine, morpholine, 4-phenyl, morpholinobenzene, 4-phenyl-morpholine, phenyl morpholine, 4-phenylmorpholin, unii-h4oc7fb1ex, h4oc7fb1ex, n-phenyl morpholine
PubChem CID 62339
IUPAC Name 4-phenylmorpholine
SMILES C1CN(CCO1)C1=CC=CC=C1

Specifications

Melting Point 54°C
Boiling Point 270°C
Quantity 25 g
UN Number 2811
Formula Weight 163.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylmorpholine
RTECSNumber QE8575000
TSCA Yes