missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylimidazole 98.0+%, TCI America™

Catalog No. p-7134823
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08775G 5 g
P087725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08775G Supplier TCI America Supplier No. P08775G
Only null left

Chemical Identifiers

CAS 670-95-1
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD00005197
InChI Key XHLKOHSAWQPOFO-UHFFFAOYSA-N
Synonym 4-phenylimidazole, 4-phenyl-1h-imidazole, 1h-imidazole, 4-phenyl, 4 5-phenylimidazole, imidazole, 4-phenyl, 4-phenyl imidazole, chembl14145, pim, 4-phenyl-1h-imidazol, 5-phenylimidazole
PubChem CID 69590
IUPAC Name 5-phenyl-1H-imidazole
SMILES C1=CC=C(C=C1)C2=CN=CN2

Specifications

Melting Point 146°C
Color White-Yellow
Quantity 5 g
Formula Weight 144.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylimidazole