accessibility menu, dialog, popup

UserName

NHN

CAS RN 670-95-1

IUPAC Name: 5-phenyl-1H-imidazole
Synonyms: 4-phenyl-3H-imidazole
Molecular Weight (g/mol): 144.18
Molecular Formula: C9H8N2
InChi Key: XHLKOHSAWQPOFO-UHFFFAOYSA-N
SMILES: N1C=NC=C1C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)