missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylimidazole, 98+%

Catalog No. AAL0223709
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0223709 10 g
AAL0223704 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0223709 Supplier Thermo Scientific Chemicals Supplier No. L0223709
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 670-95-1 | C9H8N2 | 144.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 670-95-1
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD00005197
InChI Key XHLKOHSAWQPOFO-UHFFFAOYSA-N
Synonym 4-phenylimidazole, 4-phenyl-1h-imidazole, 1h-imidazole, 4-phenyl, 4 5-phenylimidazole, imidazole, 4-phenyl, 4-phenyl imidazole, chembl14145, pim, 4-phenyl-1h-imidazol, 5-phenylimidazole
PubChem CID 69590
IUPAC Name 5-phenyl-1H-imidazole
SMILES C1=CC=C(C=C1)C2=CN=CN2

Specifications

Melting Point 143°C to 145°C
Quantity 10 g
Beilstein 2969
Formula Weight 144.18
Percent Purity ≥98%
Chemical Name or Material 4-Phenylimidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-580-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only