missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenylethynylphthalic Anhydride 98.0+%, TCI America™
$177.11 - $591.03
Chemical Identifiers
| CAS | 119389-05-8 |
|---|---|
| Molecular Formula | C16H8O3 |
| Molecular Weight (g/mol) | 248.237 |
| MDL Number | MFCD01862632 |
| InChI Key | UPGRRPUXXWPEMV-UHFFFAOYSA-N |
| PubChem CID | 11053989 |
| IUPAC Name | 5-(2-phenylethynyl)-2-benzofuran-1,3-dione |
| SMILES | C1=CC=C(C=C1)C#CC2=CC3=C(C=C2)C(=O)OC3=O |
Chemical Identifiers
| 119389-05-8 | |
| 248.237 | |
| UPGRRPUXXWPEMV-UHFFFAOYSA-N | |
| 5-(2-phenylethynyl)-2-benzofuran-1,3-dione |
| C16H8O3 | |
| MFCD01862632 | |
| 11053989 | |
| C1=CC=C(C=C1)C#CC2=CC3=C(C=C2)C(=O)OC3=O |
Specifications
| 119389-05-8 | |
| Yellow | |
| MFCD01862632 | |
| UPGRRPUXXWPEMV-UHFFFAOYSA-N | |
| 5-(2-phenylethynyl)-2-benzofuran-1,3-dione | |
| 11053989 | |
| ≥98.0% (GC,T) | |
| 4-Phenylethynylphthalic Anhydride |
| 152°C | |
| C16H8O3 | |
| 5 g | |
| C1=CC=C(C=C1)C#CC2=CC3=C(C=C2)C(=O)OC3=O | |
| 248.237 | |
| 248.24 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes