accessibility menu, dialog, popup

UserName

CAS RN 119389-05-8

IUPAC Name: 5-(2-phenylethynyl)-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 5-(2-phenylethynyl)-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 248.24
Molecular Formula: C16H8O3
InChi Key: UPGRRPUXXWPEMV-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=C1C=CC(=C2)C#CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results